Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198473.1 | NP_001185402.1 | 2 - 3 | 1053 | 80 | |
NM_106199.5 | NP_177678.2 | 2 - 3 | 1037 | 80 | ||
GenBank mRNA | AK176378.1 | - | 2 - 3 | 1037 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |