Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106217.3 | NP_565114.1 | 10 - 11 | 1396 | 96 | |
GenBank mRNA | AF286719.1 | - | 10 - 11 | 1315 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |