Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106229.2 | NP_177707.1 | 1 - 2 | 270 | 72 | |
GenBank mRNA | DQ446433.1 | - | 1 - 2 | 130 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |