Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106243.4 | NP_177721.1 | 2 - 3 | 408 | 70 | |
GenBank mRNA | AY028614.1 | - | 2 - 3 | 371 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |