Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKB2;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106304.3 | NP_177780.1 | 3 - 4 | 630 | 137 | |
GenBank mRNA | AB047279.1 | - | 3 - 4 | 431 | 137 | |
AJ297936.1 | - | 3 - 4 | 431 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |