Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CML38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198484.1 | NP_001185413.1 | 2 | 358 | 104 | |
NM_106315.3 | NP_177791.1 | 1 | 551 | 104 | ||
GenBank mRNA | BT002421.1 | - | 1 | 353 | 104 | |
BT006556.1 | - | 1 | 281 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |