Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASHH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334740.1 | NP_001321484.1 | 6 - 7 | 571 | 94 | |
NM_001334741.1 | NP_001321485.1 | 6 - 7 | 536 | 94 | ||
NM_106321.4 | NP_177797.2 | 6 - 7 | 554 | 94 | ||
NM_202429.2 | NP_974158.1 | 5 - 6 | 683 | 94 | ||
GenBank mRNA | AB493538.1 | - | 5 - 6 | 414 | 94 | |
BT001913.1 | - | 5 - 6 | 414 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |