Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106336.4 | NP_177811.2 | 3 - 4 | 807 | 88 | |
GenBank mRNA | AF479278.1 | - | 3 - 4 | 727 | 88 | |
AK118540.1 | - | 3 - 4 | 718 | 88 | ||
BT005978.1 | - | 3 - 4 | 665 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |