Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198486.2 | NP_001185415.1 | 3 - 4 | 862 | 84 | |
NM_001334757.1 | NP_001322101.1 | 3 - 4 | 737 | 84 | ||
NM_001334758.1 | NP_001322102.1 | 2 - 3 | 970 | 84 | ||
NM_106351.4 | NP_565147.1 | 2 - 3 | 970 | 84 | ||
GenBank mRNA | AY087441.1 | - | 2 - 3 | 624 | 84 | |
BT024747.1 | - | 2 - 3 | 568 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |