Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106361.4 | NP_177836.1 | 6 - 7 | 1783 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |