Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084367.2 | NP_001077836.1 | 8 - 9 | 3403 | 116 | |
NM_106379.9 | NP_177854.6 | 8 - 9 | 3555 | 116 | ||
GenBank mRNA | EU014690.1 | - | 7 - 8 | 3268 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |