Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334779.1 | NP_001320598.1 | 5 - 6 | 879 | 67 | |
NM_001334780.1 | NP_001320597.1 | 6 - 7 | 801 | 67 | ||
NM_001334781.1 | NP_001320596.1 | 3 - 4 | 375 | 67 | ||
NM_106391.2 | NP_177866.2 | 6 - 7 | 786 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |