Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334788.1 | NP_001321320.1 | 6 - 7 | 1668 | 84 | |
NM_106407.7 | NP_177882.1 | 7 - 8 | 1595 | 84 | ||
GenBank mRNA | AF503759.1 | - | 6 - 7 | 1320 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |