Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106463.3 | NP_177937.1 | 2 - 3 | 1074 | 76 | |
GenBank mRNA | AK227331.1 | - | 2064 | 76 | ||
BT004567.1 | - | 2 - 3 | 1043 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |