Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161003.2 | NP_001154475.1 | 7 - 8 | 1891 | 82 | |
NM_106474.5 | NP_177948.3 | 6 - 7 | 2006 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |