Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATGA2OX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106491.2 | NP_177965.1 | 1 - 2 | 483 | 77 | |
GenBank mRNA | AJ132435.1 | - | 1 - 2 | 413 | 77 | |
BT002763.1 | - | 1 - 2 | 373 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |