Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198498.1 | NP_001185427.1 | 6 - 7 | 1075 | 126 | |
NM_001334830.1 | NP_001322353.1 | 6 - 7 | 1173 | 126 | ||
NM_106501.5 | NP_177975.3 | 6 - 7 | 1173 | 126 | ||
GenBank mRNA | AK221532.1 | - | 6 - 7 | 1063 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |