Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334832.1 | NP_001319403.1 | 18 - 19 | 4417 | 73 | |
NM_001334833.1 | NP_001322931.1 | 17 - 18 | 4376 | 73 | ||
NM_001334834.1 | NP_001322932.1 | 17 - 18 | 4343 | 73 | ||
NM_106505.5 | NP_177979.1 | 17 - 18 | 2607 | 73 | ||
GenBank mRNA | KJ138926.1 | - | 16 - 17 | 2401 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |