Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334832.1 | NP_001319403.1 | 6 - 7 | 2999 | 89 | |
NM_001334833.1 | NP_001322931.1 | 5 - 6 | 2958 | 89 | ||
NM_001334834.1 | NP_001322932.1 | 5 - 6 | 2925 | 89 | ||
NM_106505.5 | NP_177979.1 | 5 - 6 | 1189 | 89 | ||
GenBank mRNA | KJ138926.1 | - | 4 - 5 | 983 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |