Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334861.1 | NP_001322959.1 | 9 - 10 | 1687 | 94 | |
NM_001334862.1 | NP_001322960.1 | 10 - 11 | 1453 | 94 | ||
NM_001334863.1 | NP_001322961.1 | 9 - 10 | 1611 | 94 | ||
NM_001334864.1 | NP_001322958.1 | 8 - 9 | 1181 | 94 | ||
NM_106545.4 | NP_178017.2 | 10 - 11 | 1482 | 94 | ||
GenBank mRNA | AY091131.1 | - | 10 - 11 | 1406 | 94 | |
AY114039.1 | - | 9 - 10 | 1359 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |