Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198506.1 | NP_001185435.1 | 13 - 14 | 1692 | 138 | |
NM_001198507.1 | NP_001185436.1 | 13 - 14 | 1692 | 138 | ||
NM_106578.3 | NP_178048.2 | 13 - 14 | 1654 | 138 | ||
GenBank mRNA | EF426860.1 | - | 13 - 14 | 1485 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |