Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198506.1 | NP_001185435.1 | 1 - 2 | 518 | 75 | |
NM_001198507.1 | NP_001185436.1 | 1 - 2 | 518 | 75 | ||
NM_106578.3 | NP_178048.2 | 1 - 2 | 480 | 75 | ||
GenBank mRNA | EF426860.1 | - | 1 - 2 | 311 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |