Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198506.1 | NP_001185435.1 | 41 - 42 | 4474 | 67 | |
NM_001198507.1 | NP_001185436.1 | 42 - 43 | 4537 | 67 | ||
NM_106578.3 | NP_178048.2 | 41 - 42 | 4433 | 67 | ||
GenBank mRNA | EF426860.1 | - | 41 - 42 | 4264 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |