Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106588.1 | NP_178058.1 | 3 - 4 | 2082 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |