Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106592.5 | NP_178062.1 | 10 - 11 | 1012 | 75 | |
GenBank mRNA | AF117335.1 | - | 10 - 11 | 930 | 75 | |
AF428367.1 | - | 10 - 11 | 932 | 75 | ||
AY056147.1 | - | 10 - 11 | 918 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |