Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334912.1 | NP_001320544.1 | 17 - 18 | 1985 | 75 | |
NM_106618.5 | NP_565218.1 | 16 - 17 | 2157 | 75 | ||
GenBank mRNA | AY120774.1 | - | 16 - 17 | 1873 | 75 | |
BT000142.1 | - | 16 - 17 | 1843 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |