Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036230.1 | NP_001031307.1 | 1 - 2 | 382 | 70 | |
NM_106694.4 | NP_178161.1 | 1 - 2 | 414 | 70 | ||
GenBank mRNA | AK316784.1 | - | 1 - 2 | 176 | 70 | |
AY234854.1 | - | 1 - 2 | 176 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |