Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334999.1 | NP_001319430.1 | 8 - 9 | 2717 | 99 | |
NM_001335000.1 | NP_001320420.1 | 9 - 10 | 2649 | 99 | ||
NM_001335001.1 | NP_001320419.1 | 9 - 10 | 2401 | 99 | ||
NM_106714.2 | NP_178181.1 | 9 - 10 | 2649 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |