Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106722.2 | NP_565245.1 | 16 - 17 | 2003 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |