Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPTOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111719.3 | NP_566335.1 | 6 - 7 | 1422 | 97 | |
GenBank mRNA | AY059838.1 | - | 6 - 7 | 1322 | 97 | |
AY769948.1 | - | 6 - 7 | 1322 | 97 | ||
BT008813.1 | - | 6 - 7 | 933 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |