Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATERDJ3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337788.1 | NP_001319504.1 | 1 - 2 | 203 | 60 | |
NM_111731.4 | NP_187509.1 | 1 - 2 | 229 | 60 | ||
GenBank mRNA | BT003887.1 | - | 1 - 2 | 145 | 60 | |
BT005001.1 | - | 1 - 2 | 93 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |