Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202915.1 | NP_001189844.1 | 12 - 13 | 2506 | 101 | |
NM_111744.5 | NP_566343.1 | 12 - 13 | 2751 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |