Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125130.1 | NP_001118602.1 | 2 - 3 | 492 | 99 | |
NM_001202915.1 | NP_001189844.1 | 2 - 3 | 373 | 99 | ||
NM_111744.5 | NP_566343.1 | 2 - 3 | 618 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |