Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT3G09100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111745.3 | NP_187522.2 | 10 - 11 | 1393 | 56 | |
NM_202534.2 | NP_974263.1 | 10 - 11 | 1408 | 56 | ||
GenBank mRNA | AY125498.1 | - | 10 - 11 | 1387 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |