Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT3G09100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111745.3 | NP_187522.2 | 3 - 4 | 562 | 100 | |
NM_202534.2 | NP_974263.1 | 3 - 4 | 577 | 100 | ||
GenBank mRNA | AY125498.1 | - | 3 - 4 | 556 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |