Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035587.3 | NP_001030664.1 | 7 - 8 | 637 | 86 | |
NM_111798.4 | NP_187575.2 | 6 - 7 | 720 | 86 | ||
GenBank mRNA | AK176821.1 | - | 7 - 8 | 637 | 86 | |
AK176908.1 | - | 6 - 7 | 720 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |