Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP5K9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035589.3 | NP_001030666.1 | 4 - 5 | 2062 | 112 | |
NM_001202923.1 | NP_001189852.1 | 4 - 5 | 1992 | 112 | ||
NM_001337850.1 | NP_001327647.1 | 4 - 5 | 2006 | 112 | ||
NM_111827.3 | NP_187603.1 | 3 - 4 | 2858 | 112 | ||
GenBank mRNA | AJ810853.1 | - | 3 - 4 | 1592 | 112 | |
AK229815.1 | - | 4 - 5 | 1841 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |