Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDHAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111829.4 | NP_566361.1 | 4 - 5 | 400 | 72 | |
GenBank mRNA | AY084556.1 | - | 4 - 5 | 302 | 72 | |
AY093765.1 | - | 4 - 5 | 304 | 72 | ||
BT001054.1 | - | 4 - 5 | 259 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |