Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337853.1 | NP_001326647.1 | 17 - 18 | 3794 | 66 | |
NM_111836.5 | NP_187612.5 | 16 - 17 | 3664 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |