Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337853.1 | NP_001326647.1 | 18 - 19 | 3859 | 92 | |
NM_111836.5 | NP_187612.5 | 17 - 18 | 3729 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |