Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337853.1 | NP_001326647.1 | 5 - 6 | 1794 | 65 | |
NM_111836.5 | NP_187612.5 | 4 - 5 | 1664 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |