Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RECA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111849.4 | NP_187625.2 | 3 - 4 | 450 | 77 | |
GenBank mRNA | AY072877.1 | - | 3 - 4 | 339 | 77 | |
AY136400.1 | - | 3 - 4 | 418 | 77 | ||
BT000214.1 | - | 3 - 4 | 339 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |