Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111872.6 | NP_187648.1 | 1 - 2 | 525 | 90 | |
GenBank mRNA | AY063863.1 | - | 1 - 2 | 450 | 90 | |
AY096363.1 | - | 1 - 2 | 362 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |