Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337919.1 | NP_001326206.1 | 2 - 3 | 1720 | 66 | |
NM_001337920.1 | NP_001326207.1 | 3 - 4 | 1652 | 66 | ||
NM_001337921.1 | NP_001326208.1 | 3 - 4 | 1639 | 66 | ||
NM_001337922.1 | NP_001326209.1 | 1 - 2 | 1115 | 66 | ||
NM_111959.3 | NP_187733.1 | 4 - 5 | 1513 | 66 | ||
GenBank mRNA | AF370200.1 | - | 4 - 5 | 1440 | 66 | |
AY056368.1 | - | 4 - 5 | 1417 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |