Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111987.4 | NP_187761.1 | 11 - 12 | 2004 | 113 | |
NM_202557.2 | NP_974286.1 | 6 - 7 | 1183 | 113 | ||
GenBank mRNA | AK221314.1 | - | 11 - 12 | 1730 | 113 | |
KJ138828.1 | - | 10 - 11 | 1369 | 113 | ||
U62135.1 | - | 11 - 12 | 1719 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |