Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111987.4 | NP_187761.1 | 16 - 17 | 2848 | 73 | |
NM_202557.2 | NP_974286.1 | 11 - 12 | 2027 | 73 | ||
GenBank mRNA | AK221314.1 | - | 16 - 17 | 2574 | 73 | |
KJ138828.1 | - | 15 - 16 | 2213 | 73 | ||
U62135.1 | - | 16 - 17 | 2563 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |