Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111987.4 | NP_187761.1 | 2 - 3 | 1026 | 80 | |
NM_202557.2 | NP_974286.1 | 2 - 3 | 751 | 80 | ||
GenBank mRNA | AK221314.1 | - | 2 - 3 | 752 | 80 | |
KJ138828.1 | - | 1 - 2 | 391 | 80 | ||
U62135.1 | - | 2 - 3 | 741 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |