Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111987.4 | NP_187761.1 | 6 - 7 | 1580 | 75 | |
GenBank mRNA | AK221314.1 | - | 6 - 7 | 1306 | 75 | |
KJ138828.1 | - | 5 - 6 | 945 | 75 | ||
U62135.1 | - | 6 - 7 | 1295 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |