Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338905.1 | NP_001326065.1 | 24 - 25 | 2790 | 68 | |
NM_001338906.1 | NP_001326062.1 | 24 - 25 | 2700 | 68 | ||
NM_001338907.1 | NP_001326064.1 | 24 - 25 | 2637 | 68 | ||
NM_001338908.1 | NP_001326061.1 | 23 - 24 | 2663 | 68 | ||
NM_001338909.1 | NP_001326060.1 | 24 - 25 | 2791 | 68 | ||
NM_001338910.1 | NP_001326063.1 | 24 - 25 | 2652 | 68 | ||
NM_113689.2 | NP_189410.2 | 23 - 24 | 2663 | 68 | ||
GenBank mRNA | AY822649.1 | - | 23 - 24 | 2514 | 68 | |
AY960701.1 | - | 23 - 24 | 2663 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |