Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113698.4 | NP_189420.1 | 1 - 2 | 364 | 63 | |
GenBank mRNA | AY039980.1 | - | 1 - 2 | 222 | 63 | |
AY133800.1 | - | 1 - 2 | 125 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |